tert-Butyl (3R,5S)-6-hydroxy-3,5-O-isopropylidene-3,5-dihydroxyhexanoate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
tert-Butyl (4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxane-4-acetate, 2-[(4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetic acid tert-Butyl ester, tert-Butyl 2-[(4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate, (4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxane-4-acetic acid tert-Butyl ester
CAS Number: 124655-09-0
MDL Number: MFCD23098985
Molecular Formula: C13H24O5
Molecular Weight: 260.33
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.06
Specific rotation [a]20/D: -4 deg (C=2, MeOH)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Formula:
C₁₃H₂₄O₅ MW: 260.3300 g/mol Density: 1.06 (20 °C) |
MDL Number:
MFCD23098985 CAS Number: 124655-09-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...