1-Bromo-2,4-bis(trifluoromethyl)benzene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Bis(trifluoromethyl)-1-bromobenzene

B4207-1G B4207-5G
TCB4207-1GEA 50.56 CAD
TCB4207-1G TCB4207-5G
1-Bromo-2,4-bis(trifluoromethyl)benzene ≥97.0% (by GC)
1-Bromo-2,4-bis(trifluoromethyl)benzene

CAS Number: 327-75-3
MDL Number: MFCD00074904
Molecular Formula: C8H3BrF6
Molecular Weight: 293.01
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 51
Flash Point (°C): 69
Specific Gravity (20/20): 1.75


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4-Bis(trifluoromethyl)bromobenzene

Formula: C₈H₃BrF₆
MW: 293.01 g/mol
Boiling Pt: 51 °C
Density: 1.75 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00074904
CAS Number: 327-75-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5b04dfba6ca34889dccc82685c7b101f32be51f&vid=b5b04dfba6ca34889dccc82685c7b101f32be51f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR