2,4-Bis(trifluoromethyl)benzaldehyde ≥97.0%

Supplier: TCI America

B4206-1G B4206-5G
TCB4206-1GEA 104.28 CAD
TCB4206-1G TCB4206-5G
2,4-Bis(trifluoromethyl)benzaldehyde ≥97.0%
2,4-Bis(trifluoromethyl)benzaldehyde

CAS Number: 59664-42-5
MDL Number: MFCD00083535
Molecular Formula: C9H4F6O
Molecular Weight: 242.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 68
Specific Gravity (20/20): 1.48
Storage Temperature: 0-10°C

Formula: C₉H₄F₆O
Boiling Pt: 172 °C
Density: 1.48 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00083535
CAS Number: 59664-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3a3a6d693fe26173490615890a696d943147592&vid=d3a3a6d693fe26173490615890a696d943147592&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR