(R)-1-Boc-3-Hydroxypiperidine ≥98.0%
Supplier: TCI America
Synonyms:
(R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate
CAS Number: 143900-43-0
MDL Number: MFCD04115306
Molecular Formula: C10H19NO3
Molecular Weight: 201.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 50
Flash Point (°C): 104
Specific rotation [a]20/D: -21 deg (C=5, MeOH)
Other Product Names / Synonyms: (R)-1-Boc-3-hydroxypiperidine, (R)-1-Boc-3-piperidinol
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol Melting Pt: 50 °C Flash Pt: 104 °C |
MDL Number:
MFCD04115306 CAS Number: 143900-43-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...