3-Benzyloxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B4197-25G
B4197-5G
TCB4197-25GEA
132.72
CAD
TCB4197-25G
TCB4197-5G
3-Benzyloxyaniline ≥98.0% (by GC, titration analysis)
3-Benzyloxyaniline
CAS Number: 1484-26-0
MDL Number: MFCD00007784
Molecular Formula: C13H13NO
Molecular Weight: 199.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 66
Formula:
C₁₃H₁₃NO Melting Pt: 66 °C |
MDL Number:
MFCD00007784 CAS Number: 1484-26-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...