2-Bromo-3-nitropyridine ≥98.0% (by GC)
Supplier: TCI America
B4193-1G
B4193-5G
TCB4193-1GEA
61.62
CAD
TCB4193-1G
TCB4193-5G
2-Bromo-3-nitropyridine ≥98.0% (by GC)
2-Bromo-3-nitropyridine
CAS Number: 19755-53-4
MDL Number: MFCD00955613
Molecular Formula: C5H3BrN2O2
Molecular Weight: 203.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 124
Formula:
C₅H₃BrN₂O₂ Melting Pt: 124 °C |
MDL Number:
MFCD00955613 CAS Number: 19755-53-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...