5-Bromo-2-nitrobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

B4192-1G B4192-5G
TCB4192-1GEA 33.18 CAD
TCB4192-1G TCB4192-5G
5-Bromo-2-nitrobenzaldehyde ≥98.0% (by GC)
5-Bromo-2-nitrobenzaldehyde

CAS Number: 20357-20-4
MDL Number: MFCD00456508
Molecular Formula: C7H4BrNO3
Molecular Weight: 230.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73
Lambda max.: 276 nm (CH3CN)

Formula: C₇H₄BrNO₃
Melting Pt: 73 °C
MDL Number: MFCD00456508
CAS Number: 20357-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53dc692883627a0fe76bed58ea5ffcc609d8b777&vid=53dc692883627a0fe76bed58ea5ffcc609d8b777&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR