3-Bromo-5-chlorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4191-1G B4191-5G
TCB4191-1GEA 53.72 CAD
TCB4191-1G TCB4191-5G
3-Bromo-5-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-5-chlorobenzoic acid

CAS Number: 42860-02-6
MDL Number: MFCD06797227
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 197

Formula: C₇H₄BrClO₂
Melting Pt: 197 °C
MDL Number: MFCD06797227
CAS Number: 42860-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a226e734f55eee549dc1882d76398bd6e72cbd8e&vid=a226e734f55eee549dc1882d76398bd6e72cbd8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR