3-Bromo-4-(trifluoromethoxy)aniline ≥97.0%

Supplier: TCI America

B4190-1G B4190-5G
TCB4190-1GEA 92.59 CAD
TCB4190-1G TCB4190-5G
3-Bromo-4-(trifluoromethoxy)aniline ≥97.0%
3-Bromo-4-(trifluoromethoxy)aniline

CAS Number: 191602-54-7
MDL Number: MFCD00190127
Molecular Formula: C7H5BrF3NO
Molecular Weight: 256.02
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.72

Formula: C₇H₅BrF₃NO
MW: 256.02
Density: 1.72 (20 °C)
MDL Number: MFCD00190127
CAS Number: 191602-54-7
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9ac957ec54586d96d178263f22ffbf16940d5b4&vid=a9ac957ec54586d96d178263f22ffbf16940d5b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR