Biphenyl-3-carboxaldehyde ≥96.0%

Supplier: TCI America

Synonyms: 3-Phenylbenzaldehyde, [1,1'-Biphenyl]-3-carbaldehyde

B4186-1G B4186-5G
TCB4186-1GEA 79.32 CAD
TCB4186-1G TCB4186-5G
Biphenyl-3-carboxaldehyde ≥96.0%
Biphenyl-3-carboxaldehyde

CAS Number: 1204-60-0
MDL Number: MFCD01740432
Molecular Formula: C13H10O
Molecular Weight: 182.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 110
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 3-Phenylbenzaldehyde

Formula: C₁₃H₁₀O
Boiling Pt: 125 °C
Density: 1.13 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01740432
CAS Number: 1204-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=585948b97c9fe084390d1fd0373caefec73402a0&vid=585948b97c9fe084390d1fd0373caefec73402a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR