1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulfonate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4182-25G B4182-5G
TCB4182-25GEA 742.13 CAD
TCB4182-25G TCB4182-5G
1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulfonate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-2,3-dimethylimidazolium trifluoromethanesulphonate

CAS Number: 765910-73-4
MDL Number: MFCD03427624
Molecular Formula: C10H17F3N2O3S
Molecular Weight: 302.31
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 44
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Ionic Liquids
Other Product Names / Synonyms: 1-Butyl-2,3-dimethylimidazolium Triflate

Formula: C₁₀H₁₇F₃N₂O₃S
Melting Pt: 44 °C
MDL Number: MFCD03427624
CAS Number: 765910-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfddc3802c86b38712ce2f58ae2de86d06424279&vid=cfddc3802c86b38712ce2f58ae2de86d06424279&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR