O-tert-Butyl-N,N'-diisopropylisourea ≥98.0%

Supplier: TCI America

Synonyms: tert-Butyl N,N'-diisopropylcarbamimidate, 2-tert-Butyl-1,3-diisopropylisourea

B4178-1G B4178-5G
TCB4178-1GEA 120.08 CAD
TCB4178-1G TCB4178-5G
O-tert-Butyl-N,N'-diisopropylisourea ≥98.0%
O-tert-Butyl-N,N'-diisopropylisourea

CAS Number: 71432-55-8
MDL Number: MFCD06657672
Molecular Formula: C11H24N2O
Molecular Weight: 200.33
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 61
Specific Gravity (20/20): 0.84
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification)
Other Product Names / Synonyms: 2-tert-Butyl-1,3-diisopropylisourea

Formula: C₁₁H₂₄N₂O
Boiling Pt: 61 °C
Density: 0.84 (20 °C)
MDL Number: MFCD06657672
CAS Number: 71432-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d87fa61f75f68c240e3a804da372a1f25fce1535&vid=d87fa61f75f68c240e3a804da372a1f25fce1535&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR