1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane ≥94.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane

B4167-1G B4167-5G
TCB4167-1GEA 251.22 CAD
TCB4167-1G TCB4167-5G
1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane ≥94.0% (by GC)
1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane

CAS Number: 1482416-52-3
Molecular Formula: C16BrF33O5
Molecular Weight: 979.02
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.87


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane

Formula: C₁₆BrF₃₃O₅
MW: 979.02 g/mol
Density: 1.87 (20 °C)
CAS Number: 1482416-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fa39256329b855bda960dc726c8efecdbc249c2&vid=4fa39256329b855bda960dc726c8efecdbc249c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR