4,4'-Biphenyldiboronic acid bis(pinacol)ester ≥98.0%

Supplier: TCI America

Synonyms: 4,4,5,5-Tetramethyl-2-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl}-1,3,2-dioxaborolane, 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl

B4166-1G B4166-5G
TCB4166-1GEA 172.22 CAD
TCB4166-1G TCB4166-5G
4,4'-Biphenyldiboronic acid bis(pinacol)ester ≥98.0%
4,4'-Biphenyldiboronic acid bis(pinacol) ester

CAS Number: 207611-87-8
MDL Number: MFCD08669545
Molecular Formula: C24H32B2O4
Molecular Weight: 406.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 262

Formula: C₂₄H₃₂B₂O₄
MW: 406.14 g/mol
Melting Pt: 262 °C
MDL Number: MFCD08669545
CAS Number: 207611-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9368836197c79e280ebdfd412d75ca3751590ca0&vid=9368836197c79e280ebdfd412d75ca3751590ca0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR