1-Boc-4-iodopiperidine ≥97.0% (by GC)

Supplier: TCI America

B4164-1G B4164-5G
TCB4164-1GEA 56.88 CAD
TCB4164-1G TCB4164-5G
1-Boc-4-iodopiperidine ≥97.0% (by GC)
1-Boc-4-iodopiperidine

CAS Number: 301673-14-3
MDL Number: MFCD04115041
Molecular Formula: C10H18INO2
Molecular Weight: 311.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 46
Storage Temperature: 0-10°C


Other Product Names / Synonyms: tert-Butyl 4-Iodo-1-piperidinecarboxylate, 4-Iodo-1-piperidinecarboxylic Acid tert-Butyl Ester, 1-Boc-4-iodopiperidine

Formula: C₁₀H₁₈INO₂
Melting Pt: 46 °C
MDL Number: MFCD04115041
CAS Number: 301673-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a556f2b028966777faa9f182c46ac55a06b65fb9&vid=a556f2b028966777faa9f182c46ac55a06b65fb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR