4-Bromobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4161-1G B4161-5G
TCB4161-1GEA 82.16 CAD
TCB4161-1G TCB4161-5G
4-Bromobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Bromobenzamidine hydrochloride

CAS Number: 55368-42-8
MDL Number: MFCD00464967
Molecular Formula: C7H7BrN2
Molecular Weight: 235.51
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 265

Formula: C₇H₈BrClN₂
Melting Pt: 265 °C
MDL Number: MFCD00464967
CAS Number: 55368-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c7d25fe5a1fff37b96c6d0da4a532f1bc74c3dc&vid=2c7d25fe5a1fff37b96c6d0da4a532f1bc74c3dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR