2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene ≥97.0%
Supplier: TCI America
Synonyms:
Benzo[1,2-b:4,5-b']dithiophene-2,6-diboronic Acid Bis(pinacol) Ester
B4158-1G
B4158-5G
TCB4158-1GEA
395
CAD
TCB4158-1G
76219-376
2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene ≥97.0%
2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene
CAS Number: 861398-06-3
Molecular Formula: C22H28B2O4S2
Molecular Weight: 442.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: Benzo[1,2-b:4,5-b']dithiophene-2,6-diboronic Acid Bis(pinacol) Ester
Formula:
C₂₂H₂₈B₂O₄S₂ MW: 442.20 g/mol |
CAS Number:
861398-06-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...