N-(tert-Butoxycarbonyl)glycine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Boc-Gly-OtBu, N-Boc-glycine tert-Butyl Ester

B4156-1G B4156-5G
TCB4156-1GEA 33.18 CAD
TCB4156-1G TCB4156-5G
N-(tert-Butoxycarbonyl)glycine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)glycine-tert-butyl ester

CAS Number: 111652-20-1
MDL Number: MFCD00191866
Molecular Formula: C11H21NO4
Molecular Weight: 231.29
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 67


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Boc-glycine tert-Butyl Ester, Boc-Gly-OtBu

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Melting Pt: 67 °C
MDL Number: MFCD00191866
CAS Number: 111652-20-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9aea777b1538fe071be16cf74e4110e5442a9dcf&vid=9aea777b1538fe071be16cf74e4110e5442a9dcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR