7-Bromoquinoline ≥98.0%
Supplier: TCI America
B4155-1G
B4155-5G
TCB4155-1GEA
150.1
CAD
TCB4155-1G
TCB4155-5G
7-Bromoquinoline ≥98.0%
7-Bromoquinoline
CAS Number: 4965-36-0
MDL Number: MFCD03695823
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 35
Lambda max.: 320 nm (EtOH aq.)Storage Temperature: 0-10°C
Formula:
C9H6BrN Melting Pt: 35 °C |
MDL Number:
MFCD03695823 CAS Number: 4965-36-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...