6-Bromo-m-toluic acid ≥98.0%

Supplier: TCI America

B4145-25G B4145-5G
TCB4145-25GEA 165.9 CAD
TCB4145-25G TCB4145-5G
6-Bromo-m-toluic acid ≥98.0%
6-Bromo-m-toluic acid

CAS Number: 6967-82-4
MDL Number: MFCD00079722
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: 6-Bromo-m-toluic Acid

Formula: C₈H₇BrO₂
Melting Pt: 138 °C
MDL Number: MFCD00079722
CAS Number: 6967-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45abc7c5384973f84d928d64ac75fc7bdb3f0964&vid=45abc7c5384973f84d928d64ac75fc7bdb3f0964&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR