8-Bromoisoquinoline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B4144-1G B4144-5G
TCB4144-1GEA 77.42 CAD
TCB4144-1G TCB4144-5G
8-Bromoisoquinoline ≥97.0% (by GC, titration analysis)
8-Bromoisoquinoline

CAS Number: 63927-22-0
MDL Number: MFCD04973298
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 78

Formula: C₉H₆BrN
Melting Pt: 78 °C
MDL Number: MFCD04973298
CAS Number: 63927-22-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9186e4f60b6dd7bc1463dbda545ef23bdcd4811&vid=b9186e4f60b6dd7bc1463dbda545ef23bdcd4811&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR