N-Boc-4-oxo-L-proline ≥98.0%

Supplier: TCI America

Synonyms: Boc-4-oxo-Pro-OH, (S)-1-Boc-4-oxo-2-pyrrolidinecarboxylic Acid, (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, (S)-1-(tert-Butoxycarbonyl)-4-oxo-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4-oxo-L-proline

B4141-1G B4141-5G
TCB4141-1GEA 66.36 CAD
TCB4141-1G TCB4141-5G
N-Boc-4-oxo-L-proline ≥98.0%
N-Boc-4-oxo-L-proline

CAS Number: 84348-37-8
MDL Number: MFCD01860669
Molecular Formula: C10H15NO5
Molecular Weight: 229.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 158
Specific rotation [a]20/D: 20 deg (C=0.5, Acetone)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (N-Protected), Reagents for Nitric Oxide Research
Other Product Names / Synonyms: N-Boc-4-oxo-L-proline, (S)-1-(tert-Butoxycarbonyl)-4-oxo-2-pyrrolidinecarboxylic Acid, (S)-1-Boc-4-oxo-2-pyrrolidinecarboxylic Acid

Formula: C10H15NO5
MW: 229.23 g/mol
Melting Pt: 158 °C
MDL Number: MFCD01860669
CAS Number: 84348-37-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e004bad98a64d29bdc3bf8e8c15113a128ae8e&vid=26e004bad98a64d29bdc3bf8e8c15113a128ae8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR