N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide ≥98.0%

Supplier: TCI America

Synonyms: (Tosylazanediyl)bis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), (Tosylazanediyl)bis(ethane-2,1-diyl) bis(4-methylbenzenesulfonate), N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulfonamide, N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulphonamide

B4136-25G B4136-5G
TCB4136-25GEA 268.6 CAD
TCB4136-25G TCB4136-5G
N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide ≥98.0%
N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide

CAS Number: 16695-22-0
MDL Number: MFCD00026000
Molecular Formula: C25H29NO8S3
Molecular Weight: 567.69
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 95


Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulfonamide

Formula: C₂₅H₂₉NO₈S₃
MW: 567.69 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00026000
CAS Number: 16695-22-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4014b9d3a83d13e0e121fa9d78a6444d843c2e0d&vid=4014b9d3a83d13e0e121fa9d78a6444d843c2e0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR