(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0%

Supplier: TCI America

Synonyms: (R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl Alcohol, (R)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl Alcohol, (R)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol

B4131-25G B4131-5G
TCB4131-25GEA 317.42 CAD
TCB4131-25G TCB4131-5G
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0%
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol

CAS Number: 127852-28-2
MDL Number: MFCD03093010
Molecular Formula: C10H8F6O
Molecular Weight: 258.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Specific rotation [a]20/D: 28 deg (C=1, CH3CN)
Lambda max.: 272 nm (MeOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl Alcohol, (R)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl Alcohol

Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Melting Pt: 55 °C
MDL Number: MFCD03093010
CAS Number: 127852-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364667050d879004265e134d16a3e3a8cda1fde8&vid=364667050d879004265e134d16a3e3a8cda1fde8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR