2-Bromo-4,5-diluorotoluene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4,5-Difluoro-2-methylbromobenzene, 1-Bromo-3,4-difluoro-6-methylbenzene, 1-Bromo-4,5-difluoro-2-methylbenzene
B4129-1G
B4129-5G
TCB4129-1GEA
56.25
CAD
TCB4129-1G
TCB4129-5G
2-Bromo-4,5-diluorotoluene ≥98.0% (by GC)
2-Bromo-4,5-diluorotoluene
CAS Number: 875664-38-3
MDL Number: MFCD07777160
Molecular Formula: C7H5BrF2
Molecular Weight: 207.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.62
Formula:
C₇H₅BrF₂ MW: 207.02 g/mol Density: 1.62 (20 °C) |
MDL Number:
MFCD07777160 CAS Number: 875664-38-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...