5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98.0% (by HPLC) for biochemical research

Supplier: TCI America

Synonyms: Magenta-GlcA CHA Salt, Magenta-Gluc CHA Salt, 5-Bromo-6-chloro-3-indolyl-β-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt

B4128-100MG B4128-10MG
TCB4128-100MGEA 385.2 CAD
TCB4128-100MG TCB4128-10MG
5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98.0% (by HPLC) for biochemical research
5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt

[for Biochemical Research]
CAS Number: 144110-43-0
MDL Number: MFCD00153929
Molecular Formula: C14H13BrClNO7
Molecular Weight: 521.79
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -69 deg (C=1, 50%DMF)
Storage Temperature: >-20°C


Product Categories: Bioscience, Glucuronidase Substrates
Other Product Names / Synonyms: 5-Bromo-6-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, Magenta-Gluc CHA Salt, Magenta-GlcA CHA Salt

Formula: C₁₄H₁₃BrClNO₇
MW: 521.79 g/mol
MDL Number: MFCD00153929
CAS Number: 144110-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8368106a6a5ee67d55e8baf92d6d41809217f1a&vid=a6174283fc54ec1fda2789b7f9a84ce01db9b7f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR