4-Bromothiophene-3-carbaldehyde ≥97.0%

Supplier: TCI America

B4126-1G B4126-200MG
TCB4126-1GEA 703.1 CAD
TCB4126-1G TCB4126-200MG
4-Bromothiophene-3-carbaldehyde ≥97.0%
4-Bromothiophene-3-carbaldehyde

CAS Number: 18791-78-1
MDL Number: MFCD01318960
Molecular Formula: C5H3BrOS
Molecular Weight: 191.04
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4-Bromo-3-formylthiophene

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 120 °C
MDL Number: MFCD01318960
CAS Number: 18791-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66a5d708985eac5f1a6afdb8e52cbfbecc653d2e&vid=66a5d708985eac5f1a6afdb8e52cbfbecc653d2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR