3-Bromo-2-fluoro-5-methylpyridine ≥98.0%
Supplier: TCI America
Synonyms:
3-Bromo-2-fluoro-5-picoline
B4122-1G
B4122-5G
TCB4122-1GEA
56.88
CAD
TCB4122-1G
TCB4122-5G
3-Bromo-2-fluoro-5-methylpyridine ≥98.0%
3-Bromo-2-fluoro-5-methylpyridine
CAS Number: 17282-01-8
MDL Number: MFCD03095305
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Bromo-6-fluoro-3-picoline
Formula:
C₆H₅BrFN MW: 190.01 Melting Pt: 59 °C |
MDL Number:
MFCD03095305 CAS Number: 17282-01-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...