Balsalazide disodium salt dihydrate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 5-[4-(2-Carboxyethylcarbamoyl)phenylazo]salicylic Acid Disodium Salt, Sodium (E)-5-((4-((2-carboxylatoethyl)carbamoyl)phenyl)diazenyl)-2-hydroxybenzoate dihydrate

B4121-1G B4121-5G
TCB4121-1GEA 185.18 CAD
TCB4121-1G TCB4121-5G
Balsalazide disodium salt dihydrate ≥98.0% (by HPLC, total nitrogen)
Balsalazide disodium salt dihydrate

CAS Number: 150399-21-6
MDL Number: MFCD01631117
Molecular Formula: C17H15N3O6
Molecular Weight: 401.29
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Bioscience, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 5-[4-(2-Carboxyethylcarbamoyl)phenylazo]salicylic Acid Disodium Salt

Formula: C₁₇H₁₅N₃O₆
MW: 437.31 g/mol
MDL Number: MFCD01631117
CAS Number: 150399-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9482eeefdbaf823d451167749d6eb2d003660b45&vid=9482eeefdbaf823d451167749d6eb2d003660b45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR