(S)-1,2-Butanediol ≥98.0%
Supplier: TCI America
Synonyms:
(S)-1,2-Butylene Glycol, (S)-1,2-Dihydroxybutane, (S)-Butane-1,2-diol
CAS Number: 73522-17-5
MDL Number: MFCD09953764
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 55
Specific Gravity (20/20): 1.00
Specific rotation [a]20/D: -17 deg (C=3, EtOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-1,2-Butylene Glycol, (S)-1,2-Dihydroxybutane
Formula:
C₄H₁₀O₂ Boiling Pt: 55 °C Density: 1.00 (20 °C) |
MDL Number:
MFCD09953764 CAS Number: 73522-17-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...