(R)-1,2-Butanediol ≥98.0% (by GC)

Supplier: TCI America

B4118-1G B4118-5G
TCB4118-1GEA 48.98 CAD
TCB4118-1G TCB4118-5G
(R)-1,2-Butanediol ≥98.0% (by GC)
(R)-1,2-Butanediol

CAS Number: 40348-66-1
MDL Number: MFCD09953765
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Specific Gravity (20/20): 1.00
Specific rotation [a]20/D: 17 deg (C=3, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1,2-Butylene Glycol, (R)-1,2-Dihydroxybutane

Formula: C₄H₁₀O₂
Boiling Pt: 60 °C
Density: 1.00 (20 °C)
MDL Number: MFCD09953765
CAS Number: 40348-66-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a52e1ed21f89069b11e4f94b95283bca8a7ff48f&vid=a52e1ed21f89069b11e4f94b95283bca8a7ff48f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR