(S)-(+)-1-Benzyloxy-2-propanol ≥95.0% (by GC)

Supplier: TCI America

B4117-1G B4117-5G
TCB4117-1GEA 51.19 CAD
TCB4117-1G TCB4117-5G
(S)-(+)-1-Benzyloxy-2-propanol ≥95.0% (by GC)
(S)-(+)-1-Benzyloxy-2-propanol

CAS Number: 85483-97-2
MDL Number: MFCD07367008
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 108
Specific Gravity (20/20): 1.03
Specific rotation [a]20/D: 13 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: Benzyl (S)-(+)-2-Hydroxypropyl Ether

Formula: C₁₀H₁₄O₂
Boiling Pt: 81 °C
Density: 1.03 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD07367008
CAS Number: 85483-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7174ea74074e03c93d69d4464ed950d9e4351b87&vid=7174ea74074e03c93d69d4464ed950d9e4351b87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR