6-Bromo-2,2'-bipyridyl ≥97.0%
Supplier: TCI America
Synonyms:
6-Bromo-2,2'-bipyridine
B4114-1G
B4114-5G
TCB4114-1GEA
233.84
CAD
TCB4114-1G
TCB4114-5G
6-Bromo-2,2'-bipyridyl ≥97.0%
6-Bromo-2,2'-bipyridyl
CAS Number: 10495-73-5
MDL Number: MFCD01318953
Molecular Formula: C10H7BrN2
Molecular Weight: 235.08
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 74
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 6-Bromo-2,2'-bipyridine
Formula:
C₁₀H₇BrN₂ Boiling Pt: 133 °C Melting Pt: 74 °C |
MDL Number:
MFCD01318953 CAS Number: 10495-73-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...