(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-4-(tert-Butyl)-2-(2-pyridyl)-4,5-dihydrooxazole

B4104-1G B4104-5G
TCB4104-1GEA 333.85 CAD
TCB4104-1G TCB4104-5G
(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline ≥98.0% (by GC)
(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline

CAS Number: 117408-98-7
MDL Number: MFCD22200533
Molecular Formula: C12H16N2O
Molecular Weight: 204.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Specific rotation [a]20/D: -77 deg (C=1, CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (S)-4-(tert-Butyl)-2-(2-pyridyl)-4,5-dihydrooxazole

Formula: C₁₂H₁₆N₂O
MW: 204.27 g/mol
Melting Pt: 72 °C
MDL Number: MFCD22200533
CAS Number: 117408-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de8094293daddd9ff05c3fd3a30ee7875d2b06a1&vid=de8094293daddd9ff05c3fd3a30ee7875d2b06a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR