1-(4-Bromophenyl)imidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4101-1G B4101-5G
TCB4101-1GEA 126.4 CAD
TCB4101-1G TCB4101-5G
1-(4-Bromophenyl)imidazole ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)imidazole

CAS Number: 10040-96-7
MDL Number: MFCD00060489
Molecular Formula: C9H7BrN2
Molecular Weight: 223.07
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123

Formula: C₉H₇BrN₂
Melting Pt: 123 °C
MDL Number: MFCD00060489
CAS Number: 10040-96-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f35972da9a3d07708ab53a2e449bc1ffed24b15&vid=1f35972da9a3d07708ab53a2e449bc1ffed24b15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR