Benzyl-3-oxopiperazine-1-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Benzyl 3-oxopiperazine-1-carboxylate, 3-Oxopiperazine-1-carboxylic Acid Benzyl Ester, 1-Carbobenzoxy-3-oxopiperazine, 1-Cbz-3-oxopiperazine

B4100-1G B4100-5G
TCB4100-1GEA 90.06 CAD
TCB4100-1G TCB4100-5G
Benzyl-3-oxopiperazine-1-carboxylate ≥98.0%
Benzyl-3-oxopiperazine-1-carboxylate

CAS Number: 78818-15-2
MDL Number: MFCD00173924
Molecular Formula: C12H14N2O3
Molecular Weight: 234.26
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 119


Other Product Names / Synonyms: 1-Cbz-3-oxopiperazine, 3-Oxopiperazine-1-carboxylic Acid Benzyl Ester, 1-Carbobenzoxy-3-oxopiperazine

Formula: C₁₂H₁₄N₂O₃
MW: 234.26 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00173924
CAS Number: 78818-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be1e4ac66dbaf965e3d010bcd762f1c40dbd6b4a&vid=be1e4ac66dbaf965e3d010bcd762f1c40dbd6b4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR