6-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4097-1G B4097-5G
TCB4097-1GEA 148.84 CAD
TCB4097-1G TCB4097-5G
6-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)
6-Bromo-o-toluic acid

CAS Number: 90259-31-7
MDL Number: MFCD01310788
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 109


Other Product Names / Synonyms: 6-Bromo-o-toluic Acid

Formula: C₈H₇BrO₂
Melting Pt: 109 °C
MDL Number: MFCD01310788
CAS Number: 90259-31-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ddeb5ad5c5e2ce69ba6c236bf4d414415c78ffd&vid=8ddeb5ad5c5e2ce69ba6c236bf4d414415c78ffd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR