2-(4-tert-Butylphenyl)pyridine ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-(4-(tert-Butyl)phenyl)pyridine

B4092-1G B4092-200MG
TCB4092-1GEA 851.62 CAD
TCB4092-1G TCB4092-200MG
2-(4-tert-Butylphenyl)pyridine ≥97.0% (by GC)
2-(4-tert-Butylphenyl)pyridine

CAS Number: 524713-66-4
MDL Number: MFCD07775634
Molecular Formula: C15H17N
Molecular Weight: 211.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.02

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Density: 1.02 (20 °C)
MDL Number: MFCD07775634
CAS Number: 524713-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8f0258535b60786bd3c1188cac202cad3a00806&vid=b8f0258535b60786bd3c1188cac202cad3a00806&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR