1-Butyl-3-methylimidazolium thiocyanate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B4091-25G B4091-5G
TCB4091-25GEA 322.32 CAD
TCB4091-25G TCB4091-5G
1-Butyl-3-methylimidazolium thiocyanate ≥97.0% (by titrimetric analysis)
1-Butyl-3-methylimidazolium thiocyanate

CAS Number: 344790-87-0
MDL Number: MFCD06798181
Molecular Formula: C9H15N3S
Molecular Weight: 197.30
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.07

Formula: C₉H₁₅N₃S
Density: 1.07 (20 °C)
MDL Number: MFCD06798181
CAS Number: 344790-87-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ddb9c587f14d3855049655629900a92fe9d24f1&vid=1ddb9c587f14d3855049655629900a92fe9d24f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR