2-(Benzylthio)ethanol ≥98.0% (by GC)

Supplier: TCI America

B4087-25G B4087-5G
TCB4087-25GEA 151.68 CAD
TCB4087-25G TCB4087-5G
2-(Benzylthio)ethanol ≥98.0% (by GC)
2-(Benzylthio)ethanol

CAS Number: 3878-41-9
MDL Number: MFCD00014039
Molecular Formula: C9H12OS
Molecular Weight: 168.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 2-Hydroxyethyl Benzyl Sulfide

Formula: C₉H₁₂OS
Boiling Pt: 120 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00014039
CAS Number: 3878-41-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e11d4d4ef9cf19953bedec492fe35d7eab1cd83&vid=7e11d4d4ef9cf19953bedec492fe35d7eab1cd83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR