N-(tert-Butoxycarbonyl)-1,4-phenylenediamine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
B4081-25G
B4081-5G
TCB4081-25GEA
210.14
CAD
TCB4081-25G
TCB4081-5G
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine ≥95.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine
CAS Number: 71026-66-9
MDL Number: MFCD00043022
Molecular Formula: C11H16N2O2
Molecular Weight: 208.26
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 116
Other Product Names / Synonyms: tert-Butyl (4-Aminophenyl)carbamate, (4-Aminophenyl)carbamic Acid tert-Butyl Ester, 4-(tert-Butoxycarbonylamino)aniline, 4-(Boc-amino)aniline, N-Boc-1,4-phenylenediamine
Formula:
C₁₁H₁₆N₂O₂ Melting Pt: 116 °C |
MDL Number:
MFCD00043022 CAS Number: 71026-66-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...