6-Bromo-1H-indazole ≥98.0%
Supplier: TCI America
B4080-25G
B4080-5G
B4080-1G
TCB4080-25GEA
360.4
CAD
TCB4080-25G
TCB4080-5G
TCB4080-1G
6-Bromo-1H-indazole ≥98.0%
6-Bromo-1H-indazole
CAS Number: 79762-54-2
MDL Number: MFCD03265457
Molecular Formula: C7H5BrN2
Molecular Weight: 197.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 183
Formula:
C₇H₅BrN₂ MW: 197.0300 g/mol Melting Pt: 183 °C |
MDL Number:
MFCD03265457 CAS Number: 79762-54-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...