1-Butylpiperazine ≥98.0%
Supplier: TCI America
B4078-25ML
B4078-5ML
TCB4078-25MLEA
218.2
CAD
TCB4078-25ML
TCB4078-5ML
1-Butylpiperazine ≥98.0%
1-Butylpiperazine
CAS Number: 5610-49-1
MDL Number: MFCD00039702
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Specific Gravity (20/20): 0.89
Formula:
C₈H₁₈N₂ Boiling Pt: 192 °C Density: 0.89 (20 °C) |
MDL Number:
MFCD00039702 CAS Number: 5610-49-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...