3-Bromo-2-chloro-5-nitropyridine ≥98.0% (by GC)

Supplier: TCI America

B4077-1G B4077-5G
TCB4077-1GEA 30.02 CAD
TCB4077-1G TCB4077-5G
3-Bromo-2-chloro-5-nitropyridine ≥98.0% (by GC)
3-Bromo-2-chloro-5-nitropyridine

CAS Number: 5470-17-7
MDL Number: MFCD00233989
Molecular Formula: C5H2BrClN2O2
Molecular Weight: 237.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₅H₂BrClN₂O₂
Melting Pt: 68 °C
MDL Number: MFCD00233989
CAS Number: 5470-17-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a560091e8d9a3581c8c5d555d526af335ac8b3a&vid=8a560091e8d9a3581c8c5d555d526af335ac8b3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR