N-Benzylhydroxylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4073-25G B4073-5G
TCB4073-25GEA 1107.42 CAD
TCB4073-25G TCB4073-5G
N-Benzylhydroxylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
N-Benzylhydroxylamine hydrochloride

CAS Number: 29601-98-7
MDL Number: MFCD00043377
Molecular Formula: C7H9NO
Molecular Weight: 159.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 106

Formula: C₇H₁₀ClNO
Melting Pt: 106 °C
MDL Number: MFCD00043377
CAS Number: 29601-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=901abd89b47209d3e8f283d459900f6ff399d386&vid=901abd89b47209d3e8f283d459900f6ff399d386&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR