3-tert-Butylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4072-1G B4072-5G
TCB4072-1GEA 102.7 CAD
TCB4072-1G TCB4072-5G
3-tert-Butylaniline ≥98.0% (by GC, titration analysis)
3-tert-Butylaniline

CAS Number: 5369-19-7
MDL Number: MFCD00125078
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 119
Specific Gravity (20/20): 0.95

Formula: C₁₀H₁₅N
Boiling Pt: 119 °C
Density: 0.95 (20 °C)
MDL Number: MFCD00125078
CAS Number: 5369-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e821e374a50d1e9814f44da22a052f8f5bbd76b&vid=5e821e374a50d1e9814f44da22a052f8f5bbd76b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR