5-Bromo-3-nitro-2-picoline ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-2-methyl-3-nitropyridine

B4070-1G B4070-5G
TCB4070-1GEA 33.18 CAD
TCB4070-1G TCB4070-5G
5-Bromo-3-nitro-2-picoline ≥98.0%
5-Bromo-3-nitro-2-picoline

CAS Number: 911434-05-4
MDL Number: MFCD09031419
Molecular Formula: C6H5BrN2O2
Molecular Weight: 217.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 40


Other Product Names / Synonyms: 5-Bromo-3-nitro-2-picoline

Formula: C₆H₅BrN₂O₂
Melting Pt: 40 °C
MDL Number: MFCD09031419
CAS Number: 911434-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a10754c4e986cb7ac5220821f2500b212eaf3e89&vid=a10754c4e986cb7ac5220821f2500b212eaf3e89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR