5-Bromo-2-methox-4-methylypyridine ≥98.0% (by GC)

Supplier: TCI America

B4067-1G B4067-5G
TCB4067-1GEA 44.56 CAD
TCB4067-1G TCB4067-5G
5-Bromo-2-methox-4-methylypyridine ≥98.0% (by GC)
5-Bromo-2-methox-4-methylypyridine

CAS Number: 164513-39-7
MDL Number: MFCD04039980
Molecular Formula: C7H8BrNO
Molecular Weight: 202.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 38
Flash Point (°C): 107
Storage Temperature: 0-10°C

Formula: C₇H₈BrNO
Melting Pt: 38 °C
Flash Pt: 107 °C
MDL Number: MFCD04039980
CAS Number: 164513-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d744dba62dd68967b331a4c39b5bd7b19916176a&vid=d744dba62dd68967b331a4c39b5bd7b19916176a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR