5-(2-Bromoethyl)-2,3-dihydrobenzofuran ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
5-(2-Bromoethyl)coumaran
B4066-25G
B4066-5G
TCB4066-25GEA
614.62
CAD
TCB4066-25G
TCB4066-5G
5-(2-Bromoethyl)-2,3-dihydrobenzofuran ≥98.0% (by GC)
5-(2-Bromoethyl)-2,3-dihydrobenzofuran
CAS Number: 127264-14-6
MDL Number: MFCD06797641
Molecular Formula: C10H11BrO
Molecular Weight: 227.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67
Other Product Names / Synonyms: 5-(2-Bromoethyl)coumaran
Formula:
C₁₀H₁₁BrO MW: 227.10 g/mol Melting Pt: 67 °C |
MDL Number:
MFCD06797641 CAS Number: 127264-14-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...