2-Butylimidazole-4-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America

B4064-25G B4064-5G
TCB4064-25GEA 1030.16 CAD
TCB4064-25G TCB4064-5G
2-Butylimidazole-4-carbaldehyde ≥98.0% (by GC)
2-Butylimidazole-4-carbaldehyde

CAS Number: 68282-49-5
MDL Number: MFCD01075030
Molecular Formula: C8H12N2O
Molecular Weight: 152.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 118


Other Product Names / Synonyms: 2-Butyl-5-formylimidazole, 2-Butyl-3H-imidazole-4-carboxaldehyde, 2-Butyl-4-formylimidazole

Formula: C₈H₁₂N₂O
Melting Pt: 118 °C
MDL Number: MFCD01075030
CAS Number: 68282-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7c5f0eddffb1b15b515bfc792943f173ed866f6&vid=d7c5f0eddffb1b15b515bfc792943f173ed866f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR